O-Acetylsalicylic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®

A2262-25G A2262-500G
TCA2262-25GEA 25.39 USD
TCA2262-25G TCA2262-500G
O-Acetylsalicylic acid ≥98.0%
O-Acetylsalicylic acid

CAS Number: 50-78-2
MDL Number: MFCD00002430
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Flash Point (°C): 250


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immune System, Inflammation, Allergy Research Reagents, Reagents for Inflammation & Immunity Disorder Models, Antithrombotic Ingredients, Anti-rheumatoid Arthritis Ingredients, Anti-infla
Other Product Names / Synonyms: ASA, 2-Acetoxybenzoic Acid

Formula: 2-(CH₃CO₂)C₆H₄CO₂H
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 134 °C
Density: 1.39 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002430
CAS Number: 50-78-2
EINECS: 200-064-1
Merck Index: 13,00856
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cc8faa34585dc16f5db6704f890dece94b219b6&vid=0cc8faa34585dc16f5db6704f890dece94b219b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR