6-Aminonicotinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid

A1486-1G A1486-5G
TCA1486-001GEA 42 USD
TCA1486-001G TCA1486-005G
6-Aminonicotinic acid ≥98.0% (by titrimetric analysis)
6-Aminonicotinic acid

CAS Number: 3167-49-5
MDL Number: MFCD00006326
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 312


Other Product Names / Synonyms: 6-Aminopyridine-3-carboxylic Acid

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 312 °C
MDL Number: MFCD00006326
CAS Number: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b04fea42d1a339f0657964868c3b6b8e744130c&vid=7b04fea42d1a339f0657964868c3b6b8e744130c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR