2-Amino-4-methylpyrimidine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Methylpyrimidin-2-ylamine, 4-Methylpyrimidin-2-amine

A1474-25G A1474-5G
TCA1474-25GEA 112 USD
TCA1474-25G TCA1474-005G
2-Amino-4-methylpyrimidine ≥97.0% (by titrimetric analysis)
2-Amino-4-methylpyrimidine

CAS Number: 108-52-1
MDL Number: MFCD00006101
Molecular Formula: C5H7N3
Molecular Weight: 109.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 159

Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 159 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00006101
CAS Number: 108-52-1
EINECS: 203-591-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a66cc79dffc1d3cbc558c117afc40facb7427d0&vid=3a66cc79dffc1d3cbc558c117afc40facb7427d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR