Allylchlorodimethylsilane ≥96.0%

Supplier: TCI America

Synonyms: Allyldimethylchlorosilane, ADMCS

A1275-10ML A1275-25ML
TCA1275-010MLEA 140 USD
TCA1275-010ML TCA1275-25ML
Allylchlorodimethylsilane ≥96.0%
Allylchlorodimethylsilane

[Allyldimethylsilylating Reagent for GC/MS]
CAS Number: 4028-23-3
MDL Number: MFCD00000506
Molecular Formula: C5H11ClSi
Molecular Weight: 134.68
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 5
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Silane Coupling Agents, Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Allyldimethylchlorosilane, Allyldimethylsilyl Chloride

Formula: C₅H₁₁ClSi
MW: 134.68 g/mol
Boiling Pt: 113 °C
Density: 0.89 (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00000506
CAS Number: 4028-23-3
EINECS: 223-706-2
UN: 2985
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43d0e67a25cdf6bdfdf9d0723c27cc0216c9f3ff&vid=620c439eb1d29e612584d9a68b2df34d417742c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR