1-(3-Aminopropyl)imidazole ≥97.0%
Supplier: TCI America
A1185-25G
A1185-500G
A1185-100G
TCA1185-025GEA
35.22
USD
TCA1185-025G
TCA1185-500G
TCA1185-100G
1-(3-Aminopropyl)imidazole ≥97.0%
1-(3-Aminopropyl)imidazole
CAS Number: 5036-48-6
MDL Number: MFCD00009819
Molecular Formula: C6H11N3
Molecular Weight: 125.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 154
Specific Gravity (20/20): 1.06
Formula:
C₆H₁₁N₃ Boiling Pt: 117 °C Density: 1.06 (20 °C) Flash Pt: 154 °C |
MDL Number:
MFCD00009819 CAS Number: 5036-48-6 EINECS: 225-730-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...