(±)-1-Methyl-3-phenylpropylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0999-25ML A0999-500ML
TCA0999-025MLEA 63.07 USD
TCA0999-025ML TCA0999-500ML
(±)-1-Methyl-3-phenylpropylamine ≥98.0% (by GC, titration analysis)
(±)-1-Methyl-3-phenylpropylamine

CAS Number: 22374-89-6
MDL Number: MFCD00008090
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 97
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: 1-Methyl-3-phenylpropylamine, 4-Phenyl-2-butylamine

Formula: C₁₀H₁₅N
Boiling Pt: 222 °C
Density: 0.93 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00008090
CAS Number: 22374-89-6
EINECS: 244-942-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85f3d7318e70802f4d723a3998bd1ee8cc60c1c5&vid=85f3d7318e70802f4d723a3998bd1ee8cc60c1c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR