2-Aminonicotinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

A0994-5G A0994-25G
TCA0994-005GEA 29 USD
TCA0994-005G TCA0994-025G
2-Aminonicotinic acid ≥98.0% (by titrimetric analysis)
2-Aminonicotinic acid

CAS Number: 5345-47-1
MDL Number: MFCD00006318
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Aminopyridine-3-carboxylic Acid

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 297 °C
Density: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS Number: 5345-47-1
EINECS: 226-296-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ef0ba537795ec160a717d809d9d716887ebb7a8&vid=1ef0ba537795ec160a717d809d9d716887ebb7a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR