2-Chloro-5-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America

Synonyms: 3-Amino-4-chloro-α,α,α-trifluortoluene, 3-Amino-4-chloro-α,α,α-trifluorotoluene, 6-Chloro-α,α,α-trifluoro-m-toluidine, 3-Amino-4-chlorobenzotrifluoride

A0948-500G A0948-25G
TCA0948-500GEA 374 USD
TCA0948-500G TCA0948-25G
2-Chloro-5-(trifluoromethyl)aniline ≥98.0%
2-Chloro-5-(trifluoromethyl)aniline

CAS Number: 121-50-6
MDL Number: MFCD00007673
Molecular Formula: C7H5ClF3N
Molecular Weight: 195.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Specific Gravity (20/20): 1.43


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chloro-5-trifluoromethylaniline

Formula: C₇H₅ClF₃N
Boiling Pt: 106 °C
Melting Pt: 10 °C
Density: 1.43 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007673
CAS Number: 121-50-6
EINECS: 204-475-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fafab5547588aa16e281f270c26c781c5400ee3&vid=2fafab5547588aa16e281f270c26c781c5400ee3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR