Anthranilonitrile ≥98.0%

Supplier: TCI America

A0861-500G A0861-25G A0861-100G
TCA0861-500GEA 331 USD
TCA0861-500G TCA0861-025G TCA0861-100G
Anthranilonitrile ≥98.0%
Anthranilonitrile

CAS Number: 1885-29-6
MDL Number: MFCD00007631
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 263
Melting point (°C): 50
Flash Point (°C): 145


Other Product Names / Synonyms: Anthranilonitrile, 2-Cyanoaniline

Formula: C₇H₆N₂
Boiling Pt: 263 °C
Melting Pt: 50 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00007631
CAS Number: 1885-29-6
EINECS: 217-549-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc44d50b7261adf5452d546348150268069cd413&vid=236fffe3d7d4814f3a4fddfb92bc138e038949a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR