3-Aminobenzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A0860-25G A0860-250G
TCA0860-025GEA 142 USD
TCA0860-025G TCA0860-250G
3-Aminobenzonitrile ≥98.0% (by GC, titration analysis)
3-Aminobenzonitrile

CAS Number: 2237-30-1
MDL Number: MFCD00007756
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 290
Melting point (°C): 53
Flash Point (°C): 112


Other Product Names / Synonyms: 3-Cyanoaniline

Formula: C₇H₆N₂
Boiling Pt: 290 °C
Melting Pt: 53 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007756
CAS Number: 2237-30-1
EINECS: 218-800-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=595d06677142f8ceeb3cfa7583a726dd7dca7cbb&vid=b1f5ad2031594586d5a707967a09139d60a9f37a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR