Aminoacetaldehyde dimethyl acetal ≥98.0%

Supplier: TCI America

Synonyms: 2,2-Dimethoxyethanamine

A0801-250ML A0801-25ML
TCA0801-250MLEA 276 USD
TCA0801-250ML TCA0801-25ML
Aminoacetaldehyde dimethyl acetal ≥98.0%
Aminoacetaldehyde dimethyl acetal

CAS Number: 22483-09-6
MDL Number: MFCD00008135
Molecular Formula: C4H11NO2
Molecular Weight: 105.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 53
Specific Gravity (20/20): 0.98

Formula: C₄H₁₁NO₂
Boiling Pt: 136 °C
Density: 0.98 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00008135
CAS Number: 22483-09-6
EINECS: 245-026-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eab141da540946b2a7d1e585026edbb547d9c69f&vid=eab141da540946b2a7d1e585026edbb547d9c69f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR