Acetohydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Acetic hydrazide, Acethydrazide

A0789-25G A0789-500G A0789-100G
TCA0789-025GEA 77 USD
TCA0789-025G TCA0789-500G TCA0789-100G
Acetohydrazide ≥98.0% (by GC, titration analysis)
Acetohydrazide

CAS Number: 1068-57-1
MDL Number: MFCD00007610
Molecular Formula: C2H6N2O
Molecular Weight: 74.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 66


Other Product Names / Synonyms: Acetylhydrazine

Formula: C₂H₆N₂O
MW: 74.08 g/mol
Boiling Pt: 134 °C
Melting Pt: 66 °C
Flash Pt: >100 °C
MDL Number: MFCD00007610
CAS Number: 1068-57-1
EINECS: 213-948-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1001d81aa1f20ffc26d74201284405937b84332e&vid=1001d81aa1f20ffc26d74201284405937b84332e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR