tert-Pentylbenzene ≥97.0%
Supplier: TCI America
CAS Number: 2049-95-8
MDL Number: MFCD00015187
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Flash Point (°C): 60
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: (1,1-Dimethylpropyl)benzene, 2-Methyl-2-phenylbutane
Formula:
C₆H₅C(CH₃)₂CH₂CH₃ Boiling Pt: 191 °C Density: 0.87 (20 °C) Flash Pt: 60 °C |
MDL Number:
MFCD00015187 CAS Number: 2049-95-8 EINECS: 218-076-0 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...