2-Amino-5-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline

A0732-500G A0732-25G
TCA0732-500GEA 490 USD
TCA0732-500G TCA0732-25G
2-Amino-5-methylpyridine ≥98.0%
2-Amino-5-methylpyridine

CAS Number: 1603-41-4
MDL Number: MFCD00006328
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 227
Melting point (°C): 78
Flash Point (°C): 118


Other Product Names / Synonyms: 6-Amino-3-picoline, 2-Amino-5-picoline

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 227.1
Melting Pt: 78 °C
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00006328
CAS Number: 1603-41-4
EINECS: 216-503-5
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f8fd96855ac1267308e8f3ad9518db580909436&vid=2f8fd96855ac1267308e8f3ad9518db580909436&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR