2-Amino-6-methoxybenzothiazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Methoxybenzothiazol-2-ylamine

A0715-500G A0715-25G A0715-100G
TCA0715-500GEA 150 USD
TCA0715-500G TCA0715-025G TCA0715-100G
2-Amino-6-methoxybenzothiazole ≥98.0% (by titrimetric analysis)
2-Amino-6-methoxybenzothiazole

CAS Number: 1747-60-0
MDL Number: MFCD00005787
Molecular Formula: C8H8N2OS
Molecular Weight: 180.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 168

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 168 °C
MDL Number: MFCD00005787
CAS Number: 1747-60-0
EINECS: 217-130-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2b8e1f4a1a93bc385db581ad857baaff6b7fcb5&vid=d2b8e1f4a1a93bc385db581ad857baaff6b7fcb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR