α,α,α-Trifluoro-p-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A
CAS Number: 455-14-1
MDL Number: MFCD00064396
Molecular Formula: C7H6F3N
Molecular Weight: 161.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 86
Freezing point (°C): 8
Specific Gravity (20/20): 1.30
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)aniline, alpha,alpha,alpha-Trifluoro-p-toluidine
Formula:
C₇H₆F₃N MW: 161.13 g/mol Boiling Pt: 117 °C Melting Pt: 3…8 °C Density: 1.30 (20 °C) Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064396 CAS Number: 455-14-1 EINECS: 207-236-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...