Ethyl acetoacetate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Acetoacetic acid ethyl ester, EAA

A0649-500G A0649-25G
TCA0649-500GEA 32.54 USD
TCA0649-500G TCA0649-025G
Ethyl acetoacetate ≥98.0%
Ethyl acetoacetate

CAS Number: 141-97-9
MDL Number: MFCD00009199
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Melting point (°C): -43
Flash Point (°C): 75
Specific Gravity (20/20): 1.03


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Bioscience, Lipids
Other Product Names / Synonyms: Acetoacetic Acid Ethyl Ester, Ethyl 3-Oxobutyrate, 3-Oxobutyric Acid Ethyl Ester

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8
Melting Pt: -45
Density: 1.03 (20 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44cb704dc6f8ecaaee0805da8c4000670b3bee73&vid=44cb704dc6f8ecaaee0805da8c4000670b3bee73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR