Azulene ≥99.0%
Supplier: TCI America
A0634-100MG
A0634-1G
A0634-5G
TCA0634-100MGEA
111
USD
TCA0634-100MG
TCA0634-001G
TCA0634-5G
Azulene ≥99.0%
Azulene
CAS Number: 275-51-4
MDL Number: MFCD00003810
Molecular Formula: C10H8
Molecular Weight: 128.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 242
Melting point (°C): 100
Other Product Names / Synonyms: Bicyclo[5.3.0]decapentaene, Cyclopentacycloheptene
Formula:
C₁₀H₈ Boiling Pt: 242 °C Melting Pt: 100 °C |
MDL Number:
MFCD00003810 CAS Number: 275-51-4 EINECS: 205-993-6 UN: 3077 ADR: 9,III Merck Index: 13,00928 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...