1-Aminopyrene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Pyren-1-ylamine, Pyren-1-amine
A0632-25G
A0632-5G
A0632-1G
TCA0632-25GEA
781
USD
TCA0632-25G
TCA0632-5G
TCA0632-001G
1-Aminopyrene ≥98.0% (by HPLC, titration analysis)
1-Aminopyrene
CAS Number: 1606-67-3
MDL Number: MFCD00004140
Molecular Formula: C16H11N
Molecular Weight: 217.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 118
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-Pyrenamine
Formula:
C₁₆H₁₁N MW: 217.27 g/mol Boiling Pt: 415 °C (1013 hPa) Melting Pt: 118 °C |
MDL Number:
MFCD00004140 CAS Number: 1606-67-3 EINECS: 216-521-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...