Azobenzene ≥98.0%

Supplier: TCI America
Danger

A0565-25G A0565-100G
TCA0565-25GEA 44.76 USD
TCA0565-25G TCA0565-100G
Azobenzene ≥98.0%
Azobenzene

CAS Number: 103-33-3
MDL Number: MFCD00003022
Molecular Formula: C12H10N2
Molecular Weight: 182.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 293
Melting point (°C): 68
Flash Point (°C): 100

Formula: C₁₂H₁₀N₂
MW: 182.22 g/mol
Boiling Pt: 293 °C
Melting Pt: 68 °C
Density: 1.203 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003022
CAS Number: 103-33-3
EINECS: 203-102-5
UN: 3077
ADR: 9,III
Merck Index: 12,00948
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85df272138113a000ea8b8a4ae0435a6ea13f85d&vid=85df272138113a000ea8b8a4ae0435a6ea13f85d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR