Sodium L(+)-ascorbate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt

A0539-25G A0539-500G
TCA0539-025GEA 28.2 USD
TCA0539-025G TCA0539-500G
Sodium L(+)-ascorbate ≥98.0% (by titrimetric analysis)
Sodium L(+)-ascorbate

CAS Number: 134-03-2
MDL Number: MFCD00082340
Molecular Formula: C6H8O6
Molecular Weight: 198.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 218
Specific rotation [a]20/D: 105.5 deg (C=10, H2O)


Product Categories: Bioscience, Sugar Acids, Sugars (Others), Antioxidants, Vitamins
Other Product Names / Synonyms: L-Ascorbic Acid Sodium Salt, Vitamin C Sodium Salt

Formula: C₆H₇NaO₆
MW: 198.11 g/mol
Melting Pt: 218 °C
Density: 1.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082340
CAS Number: 134-03-2
EINECS: 205-126-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67a0f6169f83c3f8d4d79739d158a6ccd9db52cc&vid=67a0f6169f83c3f8d4d79739d158a6ccd9db52cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR