Anthracene ≥97.0%

Supplier: TCI America
Warning

A0495-500G A0495-25G A0495-100G
TCA0495-500GEA 350.5 USD
TCA0495-500G TCA0495-25G TCA0495-100G
Anthracene ≥97.0%
Anthracene

CAS Number: 120-12-7
MDL Number: MFCD00001240
Molecular Formula: C14H10
Molecular Weight: 178.23
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 342
Melting point (°C): 218
Flash Point (°C): 121

Formula: C₁₄H₁₀
Boiling Pt: 342 °C
Melting Pt: 218 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00001240
CAS Number: 120-12-7
EINECS: 204-371-1
Merck Index: 13,00687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=656ea8966b19ad2eb0682e50ed47045a04da2538&vid=51b26483d28a8e3f302c2f2902892b09ae0a76aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR