p-Anisidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline
A0487-500G
A0487-25G
TCA0487-500GEA
174.61
USD
TCA0487-500G
TCA0487-25G
p-Anisidine ≥98.0% (by GC, titration analysis)
p-Anisidine
CAS Number: 104-94-9
MDL Number: MFCD00007864
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 246
Melting point (°C): 59
Flash Point (°C): 122
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Methoxyaniline
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 246 °C Melting Pt: 59 °C Density: 1.2 g/cm³ (20 °C) Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007864 CAS Number: 104-94-9 EINECS: 203-254-2 UN: 2431 ADR: 6.1,III Merck Index: 13,00670 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...