m-Anisidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-Aminoanisole, 3-Methoxyaniline, 3-Anisidine

A0485-500G A0485-25G A0485-100G
TCA0485-500GEA 329 USD
TCA0485-500G TCA0485-25G TCA0485-100G
m-Anisidine ≥98.0% (by GC, titration analysis)
m-Anisidine

CAS Number: 536-90-3
MDL Number: MFCD00007783
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 251
Flash Point (°C): 126
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-Methoxyaniline

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 251 °C
Melting Pt: –2 °C
Density: 1.10 (20 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007783
CAS Number: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2353a723fe29f691866476bd6c750e5cb3b698e6&vid=2353a723fe29f691866476bd6c750e5cb3b698e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR