Pentylamine ≥98.0%

Supplier: TCI America

A0445-500ML A0445-25ML A0445-100ML
TCA0445-500MLEA 309 USD
TCA0445-500ML TCA0445-25ML TCA0445-100ML
Pentylamine ≥98.0%
Pentylamine

CAS Number: 110-58-7
MDL Number: MFCD00008236
Molecular Formula: C5H13N
Molecular Weight: 87.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): -1
Freezing point (°C): -55
Specific Gravity (20/20): 0.75


Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction), Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminopentane, Pentylamine

Formula: H₃C(CH₂)₃CH₂NH₂
Boiling Pt: 104 °C
Melting Pt: -50 °C
Density: 0.75 (20 °C)
Flash Pt: -1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008236
CAS Number: 110-58-7
EINECS: 203-780-2
UN: 1106
ADR: 3,II
Merck Index: 13,00602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=232c71f0db99ab3d80fe4f5535d29055e7d00b2e&vid=27757a1cfd682356788de031e4d14d34bca87aed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR