(3-Aminopropyl)triethoxysilane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
3-Aminopropyltriethoxysilane, 3-(Triethoxysilyl)propylamine, Geniosil GF 93, Triethoxy(3-aminopropyl)silane
A0439-25G
A0439-100G
TCA0439-25GEA
28.49
USD
TCA0439-25G
TCA0439-100G
(3-Aminopropyl)triethoxysilane ≥98.0% (by GC, titration analysis)
(3-Aminopropyl)triethoxysilane
CAS Number: 919-30-2
MDL Number: MFCD00008207
Molecular Formula: C9H23NO3Si
Molecular Weight: 221.37
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 98
Specific Gravity (20/20): 0.95
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Triethoxy-3-aminopropylsilane, APTS
Formula:
H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃ MW: 221.37 g/mol Boiling Pt: 217 °C Melting Pt: < –70 °C Density: 0.95 (20 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008207 CAS Number: 919-30-2 EINECS: 213-048-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...