α,α,α-Trifluoro-m-toluidine ≥99.0%
Supplier: TCI America
Synonyms:
3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride
A0434-25G
A0434-100G
TCA0434-25GEA
32
USD
TCA0434-25G
TCA0434-100G
α,α,α-Trifluoro-m-toluidine ≥99.0%
α,α,α-Trifluoro-m-toluidine
CAS Number: 98-16-8
MDL Number: MFCD00007797
Molecular Formula: C7H6F3N
Molecular Weight: 161.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Melting point (°C): 3
Flash Point (°C): 85
Specific Gravity (20/20): 1.30
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Trifluoromethyl)aniline, alpha,alpha,alpha-Trifluoro-m-toluidine
Formula:
C₇H₆F₃N MW: 161.13 g/mol Boiling Pt: 189 °C Melting Pt: 3 °C Density: 1.30 (20 °C) Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007797 CAS Number: 98-16-8 EINECS: 202-643-4 UN: 2948 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...