3-Aminopyridine ≥99.0%

Supplier: TCI America
Danger

Synonyms: Pyridin-3-amine, 3-Pyridylamine

A0413-500G A0413-25G A0413-100G
TCA0413-500GEA 296 USD
TCA0413-500G TCA0413-25G TCA0413-100G
3-Aminopyridine ≥99.0%
3-Aminopyridine

CAS Number: 462-08-8
MDL Number: MFCD00006400
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 248
Melting point (°C): 62
Flash Point (°C): 88

Formula: C₅H₆N₂
Boiling Pt: 248 °C
Melting Pt: 62 °C
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006400
CAS Number: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93dba9e6da98ade269a0d230bf656616f7295555&vid=93dba9e6da98ade269a0d230bf656616f7295555&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR