2-Aminopyridine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Pyridylamine, Pyridin-2-amine
A0411-500G
A0411-25G
A0411-100G
TCA0411-500GEA
66
USD
TCA0411-500G
TCA0411-25G
TCA0411-100G
2-Aminopyridine ≥99.0% (by GC, titration analysis)
2-Aminopyridine
CAS Number: 504-29-0
MDL Number: MFCD00006312
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 61
Flash Point (°C): 68
Storage Temperature: 0-10°C
Formula:
C₅H₆N₂ MW: 94.12 g/mol Boiling Pt: 105 °C Melting Pt: 61 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006312 CAS Number: 504-29-0 EINECS: 207-988-4 UN: 2671 ADR: 6.1,II Merck Index: 13,00472 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...