2-Aminophenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Amino-1-hydroxy benzene, 2-Hydroxy aniline, 2-Hydroxyaniline, o-Aminophenol

A0335-25G A0335-100G
TCA0335-25GEA 51 USD
TCA0335-25G TCA0335-100G
2-Aminophenol ≥98.0% (by titrimetric analysis)
2-Aminophenol

CAS Number: 95-55-6
MDL Number: MFCD00007690
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 175

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.328 g/cm³ (20 °C)
Flash Pt: >175 °C
MDL Number: MFCD00007690
CAS Number: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 13,00460
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1a634727cccba28855fb83c45b6d032ee3797b1&vid=c1a634727cccba28855fb83c45b6d032ee3797b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR