Aminoguanidinium hydrogen carbonate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate
A0307-25G
A0307-500G
TCA0307-025GEA
27.47
USD
TCA0307-025G
TCA0307-500G
Aminoguanidinium hydrogen carbonate ≥99.0% (by titrimetric analysis)
Aminoguanidinium hydrogen carbonate
CAS Number: 2582-30-1
MDL Number: MFCD00012949
Molecular Formula: CH6N4
Molecular Weight: 136.11
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 162
Other Product Names / Synonyms: Aminoguanidine Hydrogen Carbonate
Formula:
C₂H₈N₄O₃ MW: 136.11 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD00012949 CAS Number: 2582-30-1 EINECS: 219-956-7 |
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...