1-(2-Aminoethyl)piperazine ≥99.0% (by GC, titration analysis)

Supplier: TCI America

A0304-500ML A0304-25ML
TCA0304-500MLEA 55.94 USD
TCA0304-500ML TCA0304-25ML
1-(2-Aminoethyl)piperazine ≥99.0% (by GC, titration analysis)
1-(2-Aminoethyl)piperazine

CAS Number: 140-31-8
MDL Number: MFCD00005971
Molecular Formula: C6H15N3
Molecular Weight: 129.21
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 222
Melting point (°C): -19
Flash Point (°C): 93
Specific Gravity (20/20): 0.99

Formula: C₆H₁₅N₃
Boiling Pt: 222 °C
Melting Pt: –19 °C
Density: 0.99 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00005971
CAS Number: 140-31-8
EINECS: 205-411-0
UN: 2815
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c589e487c6e92d3816b0d8327700217c4300aa15&vid=c589e487c6e92d3816b0d8327700217c4300aa15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR