2-(2-Aminoethylamino)ethanol ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N-(2-Hydroxyethyl)ethylenediamine, AEEA

A0299-500ML A0299-25ML
TCA0299-500MLEA 38.7 USD
TCA0299-500ML TCA0299-25ML
2-(2-Aminoethylamino)ethanol ≥99.0% (by GC, titration analysis)
2-(2-Aminoethylamino)ethanol

CAS Number: 111-41-1
MDL Number: MFCD00008170
Molecular Formula: C4H12N2O
Molecular Weight: 104.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 244
Flash Point (°C): 125
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: N-(2-Aminoethyl)ethanolamine, N-(2-Hydroxyethyl)ethylenediamine

Formula: C₄H₁₂N₂O
MW: 104.15 g/mol
Boiling Pt: 244 °C
Melting Pt: -28 °C
Density: 1.03 (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00008170
CAS Number: 111-41-1
EINECS: 203-867-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1120f1c6997fef1fd69a4bfc045c3481d9a040f1&vid=1120f1c6997fef1fd69a4bfc045c3481d9a040f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR