Aluminium isopropoxide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

A0246-500G A0246-100G
TCA0246-500GEA 41.35 USD
TCA0246-500G TCA0246-100G
Aluminium isopropoxide ≥98.0% (by titrimetric analysis)
Aluminium isopropoxide

CAS Number: 555-31-7
MDL Number: MFCD00008870
Molecular Formula: C9H21AlO3
Molecular Weight: 204.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 136

Formula: C₉H₂₁AlO₃
Boiling Pt: 135 °C
Melting Pt: 136 °C
Density: 0.9771 g/cm³ (20 °C)
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1,II
Merck Index: 13,00346
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=550038a2b7065ce12699650703bd3169a048f5aa&vid=550038a2b7065ce12699650703bd3169a048f5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR