Aluminium tri-sec-butanolate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0244-500G A0244-25G
TCA0244-500GEA 78.32 USD
TCA0244-500G TCA0244-25G
Aluminium tri-sec-butanolate ≥98.0% (by titrimetric analysis)
Aluminium tri-sec-butanolate

CAS Number: 2269-22-9
MDL Number: MFCD00009327
Molecular Formula: C12H27AlO3
Molecular Weight: 246.33
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 26


Other Product Names / Synonyms: Aluminum sec-Butylate

Formula: C₁₂H₂₇AlO₃
Boiling Pt: 206 °C
Flash Pt: 26 °C
MDL Number: MFCD00009327
CAS Number: 2269-22-9
EINECS: 218-871-2
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc593fd0731a5ccb8a61f2a98ebab8e3f396c53c&vid=bc593fd0731a5ccb8a61f2a98ebab8e3f396c53c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR