Aluminium acetylacetonate ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Tris(2,4-pentanedionato-O,O')aluminium, Acetylacetone Aluminum(III) Salt, Al(acac)3, Aluminium 2,4-pentanedionate, Aluminum(III) Acetylacetonate, Aluminium tris(2,4-pentanedionato-O,O'), Aluminum acetylacetonate, Aluminum 2,4-pentanedionate, Tris(2,4-pentanedionato)aluminum(III), Aluminum triacetylacetone
CAS Number: 13963-57-0
MDL Number: MFCD00000013
Molecular Formula: C15H21AlO6
Molecular Weight: 324.31
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 316
Melting point (°C): 195
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Acetylacetone Aluminum(III) Salt, Aluminum(III) Acetylacetonate
Formula:
C₁₅H₂₁AlO₆ MW: 324.31 g/mol Boiling Pt: 315 Melting Pt: 195 °C Density: 1.204 g/cm³ (20 °C) |
MDL Number:
MFCD00000013 CAS Number: 13963-57-0 EINECS: 237-741-6 UN: 3467 ADR: 6.1 II |
|
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...