(±)-Allyl-2,3-epoxypropyl ether ≥99.0%

Supplier: TCI America

Synonyms: Allyl glycidyl ether

A0221-500ML A0221-25ML
TCA0221-500MLEA 63.24 USD
TCA0221-500ML TCA0221-25ML
(±)-Allyl-2,3-epoxypropyl ether ≥99.0%
(±)-Allyl 2,3-epoxypropyl ether

CAS Number: 106-92-3
MDL Number: MFCD00005143
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Melting point (°C): -100
Flash Point (°C): 49
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers, Monomers
Other Product Names / Synonyms: 1-Allyl-2,3-epoxypropane

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 154 °C
Melting Pt: –100 °C
Density: 0.97 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00005143
CAS Number: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3df898c00336af12568e8b7c6d08077b578c1f19&vid=3df898c00336af12568e8b7c6d08077b578c1f19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR