Allylthiourea ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
A0220-500G
A0220-25G
A0220-100G
TCA0220-500GEA
338
USD
TCA0220-500G
TCA0220-25G
TCA0220-100G
Allylthiourea ≥98.0%
Allylthiourea
CAS Number: 109-57-9
MDL Number: MFCD00004940
Molecular Formula: C4H8N2S
Molecular Weight: 116.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 71
Product Categories: Synthetic Organic Chemistry, Thioureas (Multi-component Reaction)
Other Product Names / Synonyms: Rhodalline, Thiosinamine
Formula:
H₂CCHCH₂NHC(S)NH₂ MW: 116.19 g/mol Boiling Pt: 191 Melting Pt: 74 Density: 1.219 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004940 CAS Number: 109-57-9 EINECS: 203-683-5 UN: 2811 ADR: 6.1 III Merck Index: 12,09500 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...