D-Alanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine

A0177-25G A0177-5G
TCA0177-025GEA 84.87 USD
TCA0177-025G TCA0177-005G
D-Alanine ≥98.0% (by titrimetric analysis)
D-Alanine

CAS Number: 338-69-2
MDL Number: MFCD00008077
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -14 deg (C=2, 6mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: D-2-Aminopropionic Acid, H-D-Ala-OH

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f13b26c246f275d5c2aec68f0f98f251adb5eee9&vid=f13b26c246f275d5c2aec68f0f98f251adb5eee9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR