Adipohydrazide ≥99.0%
Supplier: TCI America
Synonyms:
Adipyl hydrazide
A0170-250G
A0170-25G
TCA0170-250GEA
116.51
USD
TCA0170-250G
TCA0170-025G
Adipohydrazide ≥99.0%
Adipohydrazide
CAS Number: 1071-93-8
MDL Number: MFCD00007614
Molecular Formula: C6H14N4O2
Molecular Weight: 174.20
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 182
Flash Point (°C): 150
Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: Adipodihydrazide
Formula:
C₆H₁₄N₄O₂ MW: 174.2 g/mol Boiling Pt: 446 Melting Pt: 182 °C Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007614 CAS Number: 1071-93-8 EINECS: 213-999-5 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...