Adipoyl dichloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A0169-500G A0169-100G A0169-25G
TCA0169-500GEA 499 USD
TCA0169-500G TCA0169-100G TCA0169-25G
Adipoyl dichloride ≥98.0% (by GC, titration analysis)
Adipoyl dichloride

CAS Number: 111-50-2
MDL Number: MFCD00000759
Molecular Formula: C6H8Cl2O2
Molecular Weight: 183.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 110
Specific Gravity (20/20): 1.26


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Adipyl Chloride

Formula: C₆H₈Cl₂O₂
Boiling Pt: 115 °C
Density: 1.26 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47fb97071e9a71049cab00f389f93e76d80dc162&vid=47fb97071e9a71049cab00f389f93e76d80dc162&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR