Bis(2-methoxyethyl)adipate ≥98.0%
Supplier: TCI America
Synonyms:
Adipic acid bis(2-methoxyethyl) ester
A0165-500ML
A0165-25ML
TCA0165-500MLEA
851
USD
TCA0165-500ML
TCA0165-025ML
Bis(2-methoxyethyl)adipate ≥98.0%
Bis(2-methoxyethyl)adipate
CAS Number: 106-00-3
MDL Number: MFCD00041905
Molecular Formula: C12H22O6
Molecular Weight: 262.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: Adipic Acid Bis(2-methoxyethyl) Ester
Formula:
C₁₂H₂₂O₆ Boiling Pt: 205 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD00041905 CAS Number: 106-00-3 EINECS: 203-352-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...