Ethyl acrylate ≥99.0% stabilized
Supplier: TCI America
Synonyms:
Acrylic acid-ethyl ester
Stabilization:
Stabilised with Stabilised
A0143-500ML
A0143-25ML
TCA0143-500MLEA
38.81
USD
TCA0143-500ML
TCA0143-25ML
Ethyl acrylate ≥99.0% stabilized
Ethyl acrylate
(stabilized with MEHQ)
CAS Number: 140-88-5
MDL Number: MFCD00009188
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Melting point (°C): -71
Flash Point (°C): 8
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Ethyl Ester
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 100 °C Melting Pt: –71 °C Density: 0.92 (20 °C) Flash Pt: 8 °C |
MDL Number:
MFCD00009188 CAS Number: 140-88-5 EINECS: 205-438-8 UN: 1917 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...