Butyl acrylate ≥99.0% stabilized

Supplier: TCI America

Synonyms: n-Butyl acrylate, Acrylic acid butyl ester

Stabilization: Stabilised with Stabilised

A0142-500ML A0142-25ML
TCA0142-500MLEA 44.82 USD
TCA0142-500ML TCA0142-25ML
Butyl acrylate ≥99.0% stabilized
Butyl acrylate

(stabilized with MEHQ)
CAS Number: 141-32-2
MDL Number: MFCD00009446
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Melting point (°C): -64
Flash Point (°C): 40
Specific Gravity (20/20): 0.90


Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Acrylic Acid Butyl Ester

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 145 °C
Melting Pt: –64 °C
Density: 0.90 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 205-480-7
UN: 2348
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=354f5004260ab2c1d7e5075cff45e6688db6d027&vid=354f5004260ab2c1d7e5075cff45e6688db6d027&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR