3-Acetylpyridine ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone

A0112-250G A0112-25G
TCA0112-250GEA 242 USD
TCA0112-250G TCA0112-25G
3-Acetylpyridine ≥98.0%
3-Acetylpyridine

CAS Number: 350-03-8
MDL Number: MFCD00006396
Molecular Formula: C7H7NO
Molecular Weight: 121.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 150
Freezing point (°C): 12
Specific Gravity (20/20): 1.11

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 220 °C
Density: 1.11 (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 12,06195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97bc34a02b78e83382805d4b92e572196b66f0e6&vid=97bc34a02b78e83382805d4b92e572196b66f0e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR