2-Acetylfuran ≥98.0%

Supplier: TCI America

Synonyms: 2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone

A0091-500G A0091-25G
TCA0091-500GEA 385 USD
TCA0091-500G TCA0091-25G
2-Acetylfuran ≥98.0%
2-Acetylfuran

CAS Number: 1192-62-7
MDL Number: MFCD00003242
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 173
Flash Point (°C): 71
Freezing point (°C): 28


Other Product Names / Synonyms: 2-Furyl Methyl Ketone

Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 173 °C
Melting Pt: 29…33 °C
Density: 1.098 g/cm³ (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00003242
CAS Number: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=881b70bfce925a7141a613a01627c72adad6ae05&vid=881b70bfce925a7141a613a01627c72adad6ae05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR