Dimethyl acetylenedicarboxylate ≥96.0%

Supplier: TCI America

Synonyms: Dimethyl but-2-yne-1,4-dioate, Dimethyl but-2-ynedioate

A0090-25ML A0090-100ML
TCA0090-25MLEA 71 USD
TCA0090-25ML TCA0090-100ML
Dimethyl acetylenedicarboxylate ≥96.0%
Dimethyl acetylenedicarboxylate

CAS Number: 762-42-5
MDL Number: MFCD00008456
Molecular Formula: C6H6O4
Molecular Weight: 142.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 86
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Acetylenedicarboxylic Acid Dimethyl Ester, DMAD

Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 198 °C
Density: 1.16 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00008456
CAS Number: 762-42-5
EINECS: 212-098-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a34cf2fde79d98d6dce2fd5e9d5dce65b78af3c0&vid=a34cf2fde79d98d6dce2fd5e9d5dce65b78af3c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR