Acetyl bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

A0080-500G A0080-25G
TCA0080-500GEA 166.52 USD
TCA0080-500G TCA0080-25G
Acetyl bromide ≥97.0% (by titrimetric analysis)
Acetyl bromide

CAS Number: 506-96-7
MDL Number: MFCD00000114
Molecular Formula: C2H3BrO
Molecular Weight: 122.95
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 77
Melting point (°C): -96
Flash Point (°C): 71
Specific Gravity (20/20): 1.65

Formula: C₂H₃BrO
MW: 122.95 g/mol
Boiling Pt: 77 °C
Melting Pt: -96 °C
Density: 1.65 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00000114
CAS Number: 506-96-7
EINECS: 208-061-7
UN: 1716
ADR: 8,II
Merck Index: 13,00084
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f5dafd2c552a4f89caa92d257c11128fa51554d&vid=9f5dafd2c552a4f89caa92d257c11128fa51554d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR