Isopropenyl acetate ≥98.0%

Supplier: TCI America

A0035-500ML A0035-25ML
TCA0035-500MLEA 54 USD
TCA0035-500ML TCA0035-025ML
Isopropenyl acetate ≥98.0%
Isopropenyl acetate

CAS Number: 108-22-5
MDL Number: MFCD00008709
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Melting point (°C): -93
Flash Point (°C): 16
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Acetic Acid Isopropenyl Ester

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 97 °C
Melting Pt: -93 °C
Density: 0.92 (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008709
CAS Number: 108-22-5
EINECS: 203-562-7
UN: 2403
ADR: 3,II
Merck Index: 13,05223
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f93b4955d0dd73ed78c3296b300aa4f2ebfc4890&vid=6aee9baa409c67e16c94ed32917c72859fc2f9f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR