tert-Butyl acetate ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1,1-Dimethylethyl acetate

A0026-500ML A0026-25ML
TCA0026-500MLEA 105.7 USD
TCA0026-500ML TCA0026-25ML
tert-Butyl acetate ≥99.0%
tert-Butyl acetate

CAS Number: 540-88-5
MDL Number: MFCD00008807
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Melting point (°C): -62
Flash Point (°C): 15
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid tert-Butyl Ester

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 96 °C
Melting Pt: –62 °C
Density: 0.87 (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00008807
CAS Number: 540-88-5
EINECS: 208-760-7
UN: 1123
ADR: 3,II
Merck Index: 13,01536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5e6003257f04841cbd20d88a45f63c6c44dc209&vid=b352bc7a2ac3671ca78fb05f2d847d25dbea7fe7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR